CAS RN 7699-45-8
Zinc bromide, anhydrous, 98%
CAS: 7699-45-8 Molecular Formula: Br2Zn Molecular Weight (g/mol): 225.188 MDL Number: MFCD00011294 InChI Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L Synonym: zincbromide,zinc ii bromide,acmc-209p7h,zinc 2+ ion dibromide,ksc378m6n,zinc bromide hydrate, puratronic™,zinc bromide, anhydrous 100g PubChem CID: 10421208 IUPAC Name: zinc;dibromide SMILES: [Zn+2].[Br-].[Br-]
Zinc bromide, 98%, extra pure, anhydrous
CAS: 7699-45-8 Molecular Formula: Br2Zn Molecular Weight (g/mol): 225.19 InChI Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L Synonym: zincbromide,zinc ii bromide,acmc-209p7h,zinc 2+ ion dibromide,ksc378m6n,zinc bromide hydrate, puratronic™,zinc bromide, anhydrous 100g PubChem CID: 10421208 IUPAC Name: zinc;dibromide SMILES: [Zn+2].[Br-].[Br-]
Zinc bromide, anhydrous, 99.999% (metals basis)
CAS: 7699-45-8 Molecular Formula: Br2Zn Molecular Weight (g/mol): 225.188 MDL Number: MFCD00011294 InChI Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L Synonym: zincbromide,zinc ii bromide,acmc-209p7h,zinc 2+ ion dibromide,ksc378m6n,zinc bromide hydrate, puratronic™,zinc bromide, anhydrous 100g PubChem CID: 10421208 IUPAC Name: zinc;dibromide SMILES: [Zn+2].[Br-].[Br-]
Zinc Bromide, Anhydrous, Reagent, 98%, Spectrum™ Chemical
CAS: 7699-45-8 Molecular Formula: Br2Zn Molecular Weight (g/mol): 225.19 InChI Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L IUPAC Name: zinc(2+) dibromide SMILES: [Zn++].[Br-].[Br-]
Zinc bromide, ultra dry, 99.999% (metals basis)
CAS: 7699-45-8 Molecular Formula: Br2Zn Molecular Weight (g/mol): 225.188 MDL Number: MFCD00011294 InChI Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L Synonym: zincbromide,zinc ii bromide,acmc-209p7h,zinc 2+ ion dibromide,ksc378m6n,zinc bromide hydrate, puratronic™,zinc bromide, anhydrous 100g PubChem CID: 10421208 IUPAC Name: zinc;dibromide SMILES: [Zn+2].[Br-].[Br-]
Zinc bromide, 99.9% (metals basis)
CAS: 7699-45-8 Molecular Formula: Br2Zn Molecular Weight (g/mol): 225.188 MDL Number: MFCD00011294 InChI Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L Synonym: zincbromide,zinc ii bromide,acmc-209p7h,zinc 2+ ion dibromide,ksc378m6n,zinc bromide hydrate, puratronic™,zinc bromide, anhydrous 100g PubChem CID: 10421208 IUPAC Name: zinc;dibromide SMILES: [Zn+2].[Br-].[Br-]
Zinc bromide, 99.999%, (trace metal basis), extra pure
CAS: 7699-45-8 Molecular Formula: Br2Zn Molecular Weight (g/mol): 225.19 MDL Number: MFCD00011294 InChI Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L Synonym: zincbromide,zinc ii bromide,acmc-209p7h,zinc 2+ ion dibromide,ksc378m6n,zinc bromide hydrate, puratronic™,zinc bromide, anhydrous 100g PubChem CID: 10421208 IUPAC Name: zinc;dibromide SMILES: [Zn+2].[Br-].[Br-]
Zinc Bromide, Anhydrous, Reagent, 98%, Spectrum™ Chemical
CAS: 7699-45-8 Molecular Formula: Br2Zn Molecular Weight (g/mol): 225.19 InChI Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L IUPAC Name: zinc(2+) dibromide SMILES: [Zn++].[Br-].[Br-]
Zinc Bromide 98.0+%, TCI America™
CAS: 7699-45-8 Molecular Formula: Br2Zn Molecular Weight (g/mol): 225.188 MDL Number: MFCD00011294 InChI Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L Synonym: zincbromide,zinc ii bromide,acmc-209p7h,zinc 2+ ion dibromide,ksc378m6n,zinc bromide hydrate, puratronic™,zinc bromide, anhydrous 100g PubChem CID: 10421208 IUPAC Name: zinc;dibromide SMILES: [Zn+2].[Br-].[Br-]
Zinc bromide, Puriss., anhydrous, ≥98%, Solstice
CAS: 7699-45-8 Molecular Formula: Br2Zn Molecular Weight (g/mol): 225.188 MDL Number: MFCD00011294 InChI Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L Synonym: zincbromide,zinc ii bromide,acmc-209p7h,zinc 2+ ion dibromide,ksc378m6n,zinc bromide hydrate, puratronic™,zinc bromide, anhydrous 100g PubChem CID: 10421208 IUPAC Name: zinc;dibromide SMILES: [Zn+2].[Br-].[Br-]
Sigma Aldrich Zinc bromide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Zinc bromide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Zinc Bromide (Crystalline/Certified), Fisher Chemical
CAS: 7699-45-8 Molecular Formula: Br2Zn Molecular Weight (g/mol): 225.188 MDL Number: MFCD00011294 InChI Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L Synonym: zincbromide,zinc ii bromide,acmc-209p7h,zinc 2+ ion dibromide,ksc378m6n,zinc bromide hydrate, puratronic™,zinc bromide, anhydrous 100g PubChem CID: 10421208 IUPAC Name: zinc;dibromide SMILES: [Zn+2].[Br-].[Br-]