Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc bromide, Puriss., anhydrous, ≥98%, Solstice

Catalog No. 60044469 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
4 x 5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-469 4 x 5 kg
60-017-73 1 kg
60-017-75 5 kg
60-017-74 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-044-469 Supplier Solstice Supplier No. 021284X5KG
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Puriss., anhydrous,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.188
MDL Number MFCD00011294
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
Synonym zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
PubChem CID 10421208
IUPAC Name zinc;dibromide
SMILES [Zn+2].[Br-].[Br-]

Specifications

Quantity 4 x 5 kg
Linear Formula ZnBr2
UN Number UN3260
Formula Weight 225.2
Percent Purity ≥98%
Grade puriss.
Chemical Name or Material Zinc Bromide
Hazard Statement P273-P280-P305 + P351 + P338-P310
Precautionary Statement H302-H314-H317-H411
DOTInformation Class 8 - PG 3 - MP - RQ - Corrosive solid, acidic, inorganic, n.o.s., HI: all
EINECSNumber 231-718-4
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.