accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO

CAS RN 77-55-4

IUPAC Name: 1-phenylcyclopentane-1-carboxylic acid
Synonyms: 1-phenylcyclopentane-1-carboxylic acid
Molecular Weight (g/mol): 190.24
Molecular Formula: C12H14O2
InChi Key: RHPCYZLXNNRRMB-UHFFFAOYSA-N
SMILES: OC(=O)C1(CCCC1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)