accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 770-35-4

IUPAC Name: 1-phenoxypropan-2-ol
Synonyms: 1-phenoxy-2-propanol 2PROPANOL1phenoxy phenoxyisopropanol 1-phenoxypropan-2-ol (8)
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: IBLKWZIFZMJLFL-UHFFFAOYNA-N
SMILES: CC(O)COC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (10)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (5)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (5)
Physical Form 
  • (7)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (5)
  • (2)
  • (3)
  • (10)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (7)
  • (1)
  • (5)
  • (2)