Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenoxy-2-propanol 97%, Thermo Scientific™

Catalog No. AC243471000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC243471000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC243471000 Supplier Thermo Scientific Chemicals Supplier No. 243471000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 770-35-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00016861
InChI Key IBLKWZIFZMJLFL-UHFFFAOYNA-N
Synonym 1-phenoxy-2-propanol, 2-propanol, 1-phenoxy, phenoxyisopropanol, propylene phenoxetol, 2-phenoxy-1-methylethanol, 1-phenoxy-propan-2-ol, phenyl-.beta.-hydroxypropyl ether, dsstox_cid_7312, dsstox_rid_78402, dsstox_gsid_27312
PubChem CID 92839
IUPAC Name 1-phenoxypropan-2-ol
SMILES CC(O)COC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.05
Infrared Spectrum Authentic
Refractive Index 1.5220 to 1.5240 (20°C, 589nm)
Linear Formula C6H5OCH2CH(OH)CH3
Quantity 100 mL
Beilstein 06, IV, 582
Formula Weight 152.19
Percent Purity ≥96.0%
Physical Form Liquid After Melting
Chemical Name or Material 1-Phenoxy-2-propanol, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.