accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7700-17-6

IUPAC Name: 1-phenylethyl (2E)-3-[(dimethoxyphosphoryl)oxy]but-2-enoate
Synonyms: crotoxyphos
Molecular Weight (g/mol): 314.27
Molecular Formula: C14H19O6P
InChi Key: XXXSILNSXNPGKG-ZHACJKMWNA-N
SMILES: COP(=O)(OC)O\C(C)=C\C(=O)OC(C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results