accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77078-22-9

IUPAC Name: 1-methoxy-2-[2-(propan-2-yloxy)ethoxy]ethane
Synonyms: 1-(2-isopropoxyethoxy)-2-methoxyethane
Molecular Weight (g/mol): 162.23
Molecular Formula: C8H18O3
InChi Key: RJBIZCOYFBKBIM-UHFFFAOYSA-N
SMILES: COCCOCCOC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results