CAS RN 771-14-2
CAS RN 771-14-2
IUPAC Name:
2-bromonaphthalen-1-amine
Synonyms:
2-bromonaphthalen-1-amine
Molecular Weight (g/mol):
222.09
Molecular Formula:
C10H8BrN
InChi Key:
YEHNFUTUJAFCAW-UHFFFAOYSA-N
SMILES:
NC1=C(Br)C=CC2=CC=CC=C12
1
–
1
of
1
results
1
1-Amino-2-bromonaphthalene 98.0+%, TCI America™
CAS: 771-14-2 Molecular Formula: C10H8BrN Molecular Weight (g/mol): 222.09 MDL Number: MFCD11870111 InChI Key: YEHNFUTUJAFCAW-UHFFFAOYSA-N Synonym: 2-Bromo-1-naphthylamine PubChem CID: 10889478 IUPAC Name: 2-bromonaphthalen-1-amine SMILES: NC1=C(Br)C=CC2=CC=CC=C12