Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-2-bromonaphthalene 98.0+%, TCI America™
SDP

Catalog No. B34115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34115G Supplier TCI America Supplier No. B34115G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 771-14-2
Molecular Formula C10H8BrN
Molecular Weight (g/mol) 222.09
MDL Number MFCD11870111
InChI Key YEHNFUTUJAFCAW-UHFFFAOYSA-N
Synonym 2-Bromo-1-naphthylamine
PubChem CID 10889478
IUPAC Name 2-bromonaphthalen-1-amine
SMILES NC1=C(Br)C=CC2=CC=CC=C12

Specifications

Melting Point 69°C
Color Red-White
Quantity 5 g
Formula Weight 222.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-2-bromonaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.