accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77134-74-8

IUPAC Name: [5-(hydroxymethyl)-1-(triphenylmethyl)-1H-imidazol-4-yl]methanol
Synonyms: [5-(hydroxymethyl)-1-(triphenylmethyl)imidazol-4-yl]methanol
Molecular Weight (g/mol): 370.45
Molecular Formula: C24H22N2O2
InChi Key: QJIJNGQOZQUGPH-UHFFFAOYSA-N
SMILES: OCC1=C(CO)N(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results