accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHNOH

CAS RN 77140-48-8

IUPAC Name: methyl 4-hydroxy-1H-indole-6-carboxylate
Synonyms: methyl 4-hydroxy-1H-indole-6-carboxylate
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: YCLHTEHHXRJHKC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(O)=C2C=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results