accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 772-03-2

CH2N

CAS RN 772-03-2

IUPAC Name: 2-ethenylquinoline
Synonyms: 2-ethenylquinoline
Molecular Weight (g/mol): 155.2
Molecular Formula: C11H9N
InChi Key: XUGNJOCQALIQFG-UHFFFAOYSA-N
SMILES: C=CC1=CC=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results