Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Vinylquinoline, Stabilized 99%, Thermo Scientific™

Catalog No. AC422600050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC422600050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC422600050 Supplier Thermo Scientific Chemicals Supplier No. 422600050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 772-03-2
Molecular Formula C11H9N
Molecular Weight (g/mol) 155.20
MDL Number MFCD00041861
InChI Key XUGNJOCQALIQFG-UHFFFAOYSA-N
Synonym 2-vinylquinoline, quinoline, 2-ethenyl, quinoline, 2-vinyl, 2-vinyl-quinoline, vinyl-quinoline, vinyl quinoline, 2-ethenyl-quinoline
PubChem CID 69875
IUPAC Name 2-ethenylquinoline
SMILES C=CC1=CC=C2C=CC=CC2=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown-Orange
Density 1.03
Infrared Spectrum Authentic
Quantity 5 g
Stabilizer 1% hydroquinone
Formula Weight 155.2
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 2-Vinylquinoline, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.