CAS RN 7720-19-6
IUPAC Name:
sodium 3-hydroxybenzoate
Synonyms:
sodium 3-hydroxybenzoate
Molecular Weight (g/mol):
160.1
Molecular Formula:
C7H5NaO3
InChi Key:
UZLYOWSQVRAOBL-UHFFFAOYSA-M
SMILES:
[Na+].OC1=CC=CC(=C1)C([O-])=O
1
–
1
of
1
results
1
Sodium 3-Hydroxybenzoate 99.0+%, TCI America™
CAS: 7720-19-6 Molecular Formula: C7H5NaO3 Molecular Weight (g/mol): 160.104 MDL Number: MFCD00058986 InChI Key: UZLYOWSQVRAOBL-UHFFFAOYSA-M Synonym: 3-Hydroxybenzoic Acid Sodium Salt PubChem CID: 23675765 IUPAC Name: sodium;3-hydroxybenzoate SMILES: C1=CC(=CC(=C1)O)C(=O)[O-].[Na+]