Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 3-Hydroxybenzoate 99.0+%, TCI America™
SDP

Catalog No. H022025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H022025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H022025G Supplier TCI America Supplier No. H022025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7720-19-6
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.104
MDL Number MFCD00058986
InChI Key UZLYOWSQVRAOBL-UHFFFAOYSA-M
Synonym 3-Hydroxybenzoic Acid Sodium Salt
PubChem CID 23675765
IUPAC Name sodium;3-hydroxybenzoate
SMILES C1=CC(=CC(=C1)O)C(=O)[O-].[Na+]

Specifications

Color Red-White
Quantity 25 g
Formula Weight 160.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-Hydroxybenzoate
RTECSNumber DH2521900
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.