accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7723-14-0

IUPAC Name: phosphorus
Synonyms: phosphorus|phosphor|phosphorus atom|phosphore|fosforo|phosphorus and metalphosphides
Molecular Weight (g/mol): 30.97
Molecular Formula: P
InChi Key: OAICVXFJPJFONN-UHFFFAOYSA-N
SMILES: [P]
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results