Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus powder, -100 mesh, red amorphous, 98% (metals basis)

Catalog No. AA1028122
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1028122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1028122 Supplier Thermo Scientific Chemicals Supplier No. 01028122

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

CAS: 7723-14-0 | P | 30.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7723-14-0
Molecular Formula P
Molecular Weight (g/mol) 30.97
MDL Number MFCD00133771
InChI Key OAICVXFJPJFONN-UHFFFAOYSA-N
Synonym red, acmc-20ajw2
PubChem CID 5462309
ChEBI CHEBI:30207
IUPAC Name phosphorus
SMILES [P]

Specifications

Color Red to Brown
Physical Form Powder
Quantity 100 g
Assay Percent Range 98.9% (metals basis)
Chemical Name or Material Phosphorus powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.