accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77237-55-9

IUPAC Name: diethyl {[4-(propan-2-yl)phenyl]methyl}phosphonate
Synonyms: diethyl (4-isopropylphenyl)methylphosphonate
Molecular Weight (g/mol): 270.31
Molecular Formula: C14H23O3P
InChi Key: IQDLJDZVTOWKTB-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=C(C=C1)C(C)C)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results