Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (4-Isopropylbenzyl)phosphonate 98.0+%, TCI America™
SDP

Catalog No. D33255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33255G Supplier TCI America Supplier No. D33255G

Chemical Identifiers

CAS 77237-55-9
Molecular Formula C14H23O3P
Molecular Weight (g/mol) 270.309
InChI Key IQDLJDZVTOWKTB-UHFFFAOYSA-N
Synonym 4-(Diethylphosphonomethyl)cumene, 1-(Diethylphosphonomethyl)-4-isopropylbenzene, (4-Isopropylbenzyl)phosphonic Acid Diethyl Ester
PubChem CID 18451666
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-propan-2-ylbenzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)C(C)C)OCC

Specifications

Color Colorless
Quantity 5 g
Formula Weight 270.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (4-Isopropylbenzyl)phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.