accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77324-87-9

CH3NOOOOCH3

CAS RN 77324-87-9

IUPAC Name: methyl 2-methyl-5-nitrobenzoate
Synonyms: methyl 2-methyl-5-nitrobenzoate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: TVGKPVDDCDQBRC-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results