Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Methyl-5-nitrobenzoate 98.0+%, TCI America™
SDP

Catalog No. M222025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M222025G 25 g
M22205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M222025G Supplier TCI America Supplier No. M222025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 77324-87-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
InChI Key TVGKPVDDCDQBRC-UHFFFAOYSA-N
Synonym methyl 5-nitro-2-methylbenzoate, 2-methyl-5-nitro-benzoic acid methyl ester, 2-methyl-5-nitrobenzoic acid methyl ester, methyl2-methyl-5-nitrobenzoate, methyl 3-nitro-6-methylbenzoate, benzoic acid, 2-methyl-5-nitro-, methyl ester, methyl 2-methyl-5-nitro-benzoate, acmc-1bix6, methyl 5-nitro-o-toluate, ksc495s6b
PubChem CID 4377758
IUPAC Name methyl 2-methyl-5-nitrobenzoate
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 69°C
Color Yellow
Quantity 25 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Methyl-5-nitrobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.