accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOH

CAS RN 7735-42-4

IUPAC Name: hexadecane-1,16-diol
Synonyms: hexadecane-1,16-diol
Molecular Weight (g/mol): 258.45
Molecular Formula: C16H34O2
InChi Key: GJBXIPOYHVMPQJ-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCCCCCCCO
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)