Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,16-Hexadecanediol 95.0+%, TCI America™
SDP

Catalog No. H05525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05525G 5 g
H05521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05525G Supplier TCI America Supplier No. H05525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7735-42-4
Molecular Formula C16H34O2
Molecular Weight (g/mol) 258.45
MDL Number MFCD00002821
InChI Key GJBXIPOYHVMPQJ-UHFFFAOYSA-N
Synonym 1,16-Dihydroxyhexadecane, Hexadecamethylene Glycol
PubChem CID 82184
IUPAC Name hexadecane-1,16-diol
SMILES OCCCCCCCCCCCCCCCCO

Specifications

Melting Point 92°C
Color White
Boiling Point 200°C
Quantity 5 g
Formula Weight 258.45
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,16-Hexadecanediol
EINECSNumber (2)-0240
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.