accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 774-81-2

IUPAC Name: 2-(3-bromo-4-methoxyphenyl)acetic acid
Synonyms: (3-bromo-4-methoxyphenyl)acetic acid
Molecular Weight (g/mol): 245.07
Molecular Formula: C9H9BrO3
InChi Key: POTVGQUUEQTPNA-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(O)=O)C=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results