accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClN

CAS RN 7742-73-6

IUPAC Name: 1,3-dichloroisoquinoline
Synonyms: 1,3-dichloroisoquinoline
Molecular Weight (g/mol): 198.05
Molecular Formula: C9H5Cl2N
InChi Key: BRGZEQXWZWBPJH-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)