CAS RN 77447-40-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Conductivity Standards for METTLER TOLEDO Conductivity Probes, METTLER TOLEDO™
Recommended NIST traceable solutions for calibrating or verifying METTLER TOLEDO conductivity electrodes.