accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNCH2OHCl

CAS RN 77481-82-4

IUPAC Name: 1-benzyl-5-ethenyl-2-[hydroxy(6-methoxyquinolin-4-yl)methyl]-1-azabicyclo[2.2.2]octan-1-ium chloride
Synonyms: 1-benzyl-5-ethenyl-2-[hydroxy(6-methoxyquinolin-4-yl)methyl]-1-azabicyclo[2.2.2]octan-1-ium chloride
Molecular Weight (g/mol): 451.01
Molecular Formula: C27H31ClN2O2
InChi Key: JYDIJFKNXHPWBJ-UHFFFAOYNA-M
SMILES: [Cl-].COC1=CC=C2N=CC=C(C(O)C3CC4CC[N+]3(CC3=CC=CC=C3)CC4C=C)C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results