Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylquinidinium Chloride 98.0+%, TCI America™
SDP

Catalog No. B16845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B16845G Supplier TCI America Supplier No. B16845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 77481-82-4
Molecular Formula C27H31ClN2O2
Molecular Weight (g/mol) 451.007
MDL Number MFCD00198106
InChI Key JYDIJFKNXHPWBJ-SRSULIGWSA-M
PubChem CID 131859232
IUPAC Name (S)-[(1S,2R,4S,5R)-1-benzyl-5-ethenyl-1-azoniabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;chloride
SMILES COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CC[N+]3(CC4C=C)CC5=CC=CC=C5)O.[Cl-]

Specifications

Quantity 5 g
Formula Weight 451.01
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylquinidinium Chloride [Chiral Phase-Transfer Catalyst]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.