CAS RN 77482-44-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ 2-Guanidinoethylmercaptosuccinic Acid
Potent inhibitor of a carboxypeptidase B-like processing enzyme referred to as enkephalin convertase (Ki = 8.8 nM).