Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 2-Guanidinoethylmercaptosuccinic Acid

Catalog No. 3693345MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-933-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-933-45MG Supplier MilliporeSigma™ Supplier No. 3693345MG
Add to Cart
Add to Cart

Potent inhibitor of a carboxypeptidase B-like processing enzyme referred to as enkephalin convertase (Ki = 8.8 nM).

Potent inhibitor of a carboxypeptidase B-like processing enzyme referred to as enkephalin convertase (Ki = 8.8 nM). Ideal for use in affinity chromatography of the enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by TLC
Target Proteins Enkephalin convertase
CAS 77482-44-1
Formula Weight 235.2g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.