accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 775-06-4

IUPAC Name: 2-amino-3-(3-hydroxyphenyl)propanoic acid
Synonyms: m-L-tyrosine|L-m-tyrosine|m-tyrosine, L-|m-tyrosine, DL-|DL-3-tyrosine
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: JZKXXXDKRQWDET-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC(O)=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)