Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3-Hydroxyphenyl)-DL-alanine, 98%, Thermo Scientific Chemicals

Catalog No. AAH6316306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6316306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6316306 Supplier Thermo Scientific Chemicals Supplier No. H6316306

missing translation for 'validMainframeMsg'

CAS: 775-06-4 | Molecular Formula: C9H11NO3 | Molecular Weight: 181.19 g/mol

3-(3-Hydroxyphenyl)-DL-alanine is used as a food and feed additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Hydroxyphenyl)-DL-alanine is used as a food and feed additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 1M HCl.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 775-06-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002597
InChI Key JZKXXXDKRQWDET-UHFFFAOYNA-N
Synonym dl-m-tyrosine, 2-amino-3-3-hydroxyphenyl propanoic acid, 3-3-hydroxyphenyl-dl-alanine, m-tyrosine, dl, d,l-metatyrosine, 3-hydroxy-dl-phenylalanine, dl-meta-tyrosine, dl-phenylalanine, 3-hydroxy, 3-m-hydroxyphenyl-dl-alanine, dl-3-tyrosine
PubChem CID 13052
IUPAC Name 2-amino-3-(3-hydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=CC(O)=C1)C(O)=O

Specifications

Melting Point 280°C to 285°C (decomposition)
Color White to Yellow
Beilstein 2416853
Merck Index 14,9840
Solubility Information Soluble in 1M HCl.
Quantity 5 g
Formula Weight 181.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-(3-Hydroxyphenyl)-DL-alanine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-270-9
RTECSNumber YP2278000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.