CAS RN 775-15-5
CAS RN 775-15-5
IUPAC Name:
1-benzylpyrrolidin-3-ol
Synonyms:
1-benzylpyrrolidin-3-ol
Molecular Weight (g/mol):
177.25
Molecular Formula:
C11H15NO
InChi Key:
YQMXOIAIYXXXEE-UHFFFAOYNA-N
SMILES:
OC1CCN(CC2=CC=CC=C2)C1
1
–
1
of
1
results
1
1-Benzyl-3-pyrrolidinol 96.0+%, TCI America™
CAS: 775-15-5 Molecular Formula: C11H15NO Molecular Weight (g/mol): 177.247 MDL Number: MFCD00012132 InChI Key: YQMXOIAIYXXXEE-UHFFFAOYSA-N PubChem CID: 69889 IUPAC Name: 1-benzylpyrrolidin-3-ol SMILES: C1CN(CC1O)CC2=CC=CC=C2