accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSiH3COCH3

CAS RN 775-56-4

IUPAC Name: diethoxy(methyl)phenylsilane
Synonyms: silane, diethoxymethylphenyl-
Molecular Weight (g/mol): 210.35
Molecular Formula: C11H18O2Si
InChi Key: MNFGEHQPOWJJBH-UHFFFAOYSA-N
SMILES: CCO[Si](C)(OCC)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)