Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl (Boc-Amino)Malonate

Catalog No. 44848625ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-848-625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 44-848-625ML Supplier Sigma Aldrich Supplier No. 44848625ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethyl (Boc-Amino)Malonate

Specifications

CAS 775-56-4
Density 0.963 g/mL at 25°C
Boiling Point 117°C (31 mmHg, lit.)
Molecular Formula C11H18O2Si
Refractive Index n20/D 1.438
Linear Formula C6H5Si(OC2H5)2CH3
MDL Number MFCD00048541
Quantity 25 mL
Synonym (Boc-amino)malonic acid diethyl ester; Diethyl 2-[N-(tert-butoxycarbonyl)amino]malonate
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.