CAS RN 775294-82-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Polo-like Kinase Inhibitor IV, SBE13
The Polo-like Kinase Inhibitor IV, SBE13, also referenced under CAS 775294-82-1, controls the biological activity of Polo-like Kinase. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.