CAS RN 7757-82-6
Sodium Sulfate Anhydrous, Certified ACS, Suitable for Pesticide Residue Analysis (Granular, 10-60 mesh), Fisher Chemical™
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L Synonym: sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 PubChem CID: 24436 ChEBI: CHEBI:32149 IUPAC Name: disodium sulfate SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sodium Sulfate Anhydrous (Low Nitrogen), Fisher Chemical™
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L Synonym: sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 PubChem CID: 24436 ChEBI: CHEBI:32149 IUPAC Name: disodium sulfate SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sodium Sulfate Anhydrous (Powder/USP/FCC), Fisher Chemical™
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L Synonym: sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 PubChem CID: 24436 ChEBI: CHEBI:32149 IUPAC Name: disodium sulfate SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sulfate Standard, 1000 ppm SO42-, Ricca Chemical
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L PubChem CID: 24436 ChEBI: CHEBI:32149 IUPAC Name: disodium sulfate SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sodium sulfate, anhydrous, 99%
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L Synonym: sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 PubChem CID: 24436 ChEBI: CHEBI:32149 SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sodium sulfate, 99%, extra pure, anhydrous
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L Synonym: sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 PubChem CID: 24436 ChEBI: CHEBI:32149 SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sodium Sulfate Anhydrous (Granular/Certified ACS), Fisher Chemical™
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L Synonym: sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 PubChem CID: 24436 ChEBI: CHEBI:32149 IUPAC Name: disodium sulfate SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sodium sulfate, 99+%, ACS reagent, granular, anhydrous
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L Synonym: sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 PubChem CID: 24436 ChEBI: CHEBI:32149 SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sodium Sulfate, Anhydrous , 99.0% min., ACS, MilliporeSigma™
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L Synonym: sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 PubChem CID: 24436 ChEBI: CHEBI:32149 IUPAC Name: disodium sulfate SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sulfate Standard, Certified, 100ppm ±1ppm (1mL = 0.1mg SO4), LabChem™
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L PubChem CID: 24436 ChEBI: CHEBI:32149 IUPAC Name: disodium sulfate SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sodium Sulfate, Anhydrous, 99.0% min., ACS, MilliporeSigma™
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L Synonym: sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 PubChem CID: 24436 ChEBI: CHEBI:32149 IUPAC Name: disodium sulfate SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sodium sulfate, 99+%, ACS reagent, anhydrous
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L Synonym: sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 PubChem CID: 24436 ChEBI: CHEBI:32149 IUPAC Name: disodium sulfate SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sodium sulfate, 99+%, for analysis, powder, anhydrous
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L Synonym: sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1 PubChem CID: 24436 ChEBI: CHEBI:32149 SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sulfate Standard, Certified, 1000ppm ±10ppm (1mL = 1mg SO4), LabChem™
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04 MDL Number: MFCD00003504 InChI Key: PMZURENOXWZQFD-UHFFFAOYSA-L PubChem CID: 24436 ChEBI: CHEBI:32149 IUPAC Name: disodium sulfate SMILES: [Na+].[Na+].[O-]S([O-])(=O)=O
Sodium Sulfate Anhydrous Granulated, 99.0%, MilliporeSigma™
CAS: 7757-82-6 Molecular Formula: Na2O4S Molecular Weight (g/mol): 142.04