Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate Anhydrous Granulated, 99.0%, MilliporeSigma™

Catalog No. M1066390500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1066390500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1066390500 Supplier MilliporeSigma Supplier No. 1.06639.0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium sulfate anhydrous granulated for organic trace analysis EMSURE™.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04

Specifications

Melting Point 888°C
pH 5.2 to 8.0 (50 g/L, H2O, 20°C)
Physical Form Granules
Quantity 500 g
Assay Percent Range ≥ 99.0%
Solubility Information 200 g/L
Percent Purity ≥99.0%
Grade Not Available
Packaging Glass Bottle
Density 2.70 g/cm3 (at 20°C); Bulk Density: 1400 to 1600 kg/m3
Chemical Name or Material Sodium Sulfate
Show More Show Less
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.