accessibility menu, dialog, popup

UserName

Viewing profile as user:
KOBrOO

CAS RN 7758-01-2

IUPAC Name: potassium bromate
Synonyms: potassium bromate bromic acid, potassium salt potassium trioxobromate
Molecular Weight (g/mol): 167
Molecular Formula: BrKO3
InChi Key: OCATYIAKPYKMPG-UHFFFAOYSA-M
SMILES: [K+].[O-][Br](=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (7)
Grade 
  • (4)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (2)
  • (7)
  • (1)
  • (3)
  • (7)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)