Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromate Standard, 1000 ppm BrO3- (with Ethylenediamine preservative), Ricca Chemical
SDP

Catalog No. 116032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1160-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1160-32 Supplier Ricca Chemical Company Supplier No. 116032
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7758-01-2 , 7732-18-5 , 7758-01-2
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]

Specifications

Packaging Amber Poly Bottle
CAS Min % 97.0, 0.87, 0.13
CAS Max % 100.0, 0.91, 0.13
Equivalence 1 mL = 1 mg BrO
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 1 L
Grade Laboratory
Chemical Name or Material Bromate Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.