accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7758-23-8

IUPAC Name: calcium didihydrogen phosphate
Synonyms: monocalcium phosphate calcium bis(dihydrogenphosphate)
Molecular Weight (g/mol): 234.05
Molecular Formula: CaH4O8P2
InChi Key: YYRMJZQKEFZXMX-UHFFFAOYSA-L
SMILES: [Ca++].OP(O)([O-])=O.OP(O)([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)
Grade 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)