Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate, Monobasic, Monohydrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02002541 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-541 500 g Poly Bottle
02-002-540 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-002-541 Supplier Avantor Supplier No. 142601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7758-23-8
Molecular Formula CaH4O8P2
Molecular Weight (g/mol) 234.05
InChI Key YYRMJZQKEFZXMX-UHFFFAOYSA-L
Synonym calcium biphosphate, monobasic calcium phosphate, calcium dihydrogen phosphate, acid calcium phosphate, primary calcium phosphate, monocalcium orthophosphate, c 38 phosphate, calcium diorthophosphate, monocalcium phosphate, monobasic, calcium dihydrogen orthophosphate
PubChem CID 24454
ChEBI CHEBI:35433
IUPAC Name calcium;dihydrogen phosphate
SMILES OP(=O)(O)[O-].OP(=O)(O)[O-].[Ca+2]

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 252.07
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.22kg
Chemical Name or Material Calcium Phosphate, Monobasic, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.