accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NSOONHON

CAS RN 7758-79-4

IUPAC Name: 4-amino-N-(1,2-oxazol-5-yl)benzene-1-sulfonamide
Synonyms: 4-amino-N-(1,2-oxazol-5-yl)benzenesulfonamide
Molecular Weight (g/mol): 239.25
Molecular Formula: C9H9N3O3S
InChi Key: CVLLLVDWOLWLPK-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results