Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Phosphate Dibasic Anhydrous
SCI_ED

Catalog No. S25563 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Reagent Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25563 100 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25563
Add to Cart
Add to Cart

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 7758-79-4
Molecular Formula C9H9N3O3S
Molecular Weight (g/mol) 239.249
InChI Key CVLLLVDWOLWLPK-UHFFFAOYSA-N
Synonym n-isoxazol-5-yl sulphanilamide, benzenesulfonamide, 4-amino-n-5-isoxazolyl, 4-amino-n-isoxazol-5-yl benzenesulfonamide, 4-amino-n-1,2-oxazol-5-yl benzenesulfonamide, sodium phosphate dibasic anhyd acs, n-isoxazol-5-yl sulfanilamide, benzenesulfonamide,4-amino-n-5-isoxazolyl, 4-amino-n-5-isoxazolylbenzenesulfonamide
PubChem CID 82193
IUPAC Name 4-amino-N-(1,2-oxazol-5-yl)benzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)NC2=CC=NO2

Specifications

Physical Form Solid
Quantity 100 g
Assay Percent Range >99%
Linear Formula Na2HPO4
Formula Weight 141.96
Packaging Reagent Grade
Chemical Name or Material Sodium Phosphate, Dibasic, Anhydrous
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.