CAS RN 7758-89-6
Copper(I) chloride, 99.99%, (trace metal basis), extra pure
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 99 MDL Number: MFCD00010971 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: chlorocopper SMILES: Cl[Cu]
Copper(I) chloride, 97%
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 98.996 MDL Number: MFCD00010971 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: chlorocopper SMILES: Cl[Cu]
Copper(I) chloride, 97%, pure, with anti-caking agent
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 99 MDL Number: MFCD00010971 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: chlorocopper SMILES: Cl[Cu]
Copper(I) Chloride, Solstice
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 98.996 MDL Number: MFCD00010971 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: chlorocopper SMILES: Cl[Cu]
Copper(I) chloride, 90+%, ACS reagent
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 99 MDL Number: MFCD00010971 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: chlorocopper SMILES: Cl[Cu]
Copper(I) chloride, 99.999% (metals basis)
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 98.996 MDL Number: MFCD00010971 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: chlorocopper SMILES: Cl[Cu]
Copper(I) chloride, 99% (metals basis)
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 98.996 MDL Number: MFCD00010971 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: chlorocopper SMILES: Cl[Cu]
Copper(I) chloride, 99%, extra pure, purified
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 99 MDL Number: MFCD00010971 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: chlorocopper SMILES: Cl[Cu]
Cuprous Chloride, ≥90%, (ACS Reagent Grade), MP Biomedicals
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 98.996 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: chlorocopper SMILES: Cl[Cu]
Copper(I) chloride, 97+%, extra pure
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 99 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: chlorocopper SMILES: Cl[Cu]
Cuprous Chloride Reagent, Acid, Ricca Chemical
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 99.00 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: λ1-copper(1+) chloride SMILES: [Cl-].[Cu+]
Cuprous Chloride Reagent, for Cyanide Determination, Ricca Chemical
CAS: 7758-89-6 Molecular Formula: ClCu Molecular Weight (g/mol): 99.00 InChI Key: OXBLHERUFWYNTN-UHFFFAOYSA-M Synonym: cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech PubChem CID: 62652 ChEBI: CHEBI:53472 IUPAC Name: λ1-copper(1+) chloride SMILES: [Cl-].[Cu+]
Sigma Aldrich Copper(I) chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Copper(I) chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2