Learn More
Copper(I) chloride, 99% (metals basis)
Description
Copper(I) chloride is precursor to many copper compounds including copper oxychloride and many organocuprate compounds of synthetic interest. It is used as a catalyst as well as a reagent in many organic reactions including Gatterman-Koch, Sandmeyer, Grignard and Gilman reactions. It catalyzes 1,4-addition of Grignard reagents to alpha,beta-unsaturated ketones. It is also used as a doping salt to enhance the cationization of polystyrene in matrix-assisted laser desorption/ionization mass spectrometry. It is also an intermediate formed in the Wacker process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1490°C |
| Melting Point | 430°C |
| Physical Form | Powder |
| Quantity | 500 g |
| Refractive Index | 1.93 |
| Sensitivity | Air and moisture sensitive |
| Appearance | Powder |
| Assay Percent Range | 99% (Metal basis) |
| Density | 4.14 |
| Solubility Information | Very soluble in concentrated hydrochloric acid. Soluble in ammonium hydroxide. Insoluble in ethanol and acetone. Sparingly soluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H302+H312-H315-H318 |
| Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c |
| DOTInformation | DOT Class: 8, Packing Group: III |
| EINECSNumber | 231-842-9 |
| RTECSNumber | GL6990000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.