accessibility menu, dialog, popup

UserName

Viewing profile as user:
PbOCrOOO

CAS RN 7758-97-6

IUPAC Name: λ²-lead(2+) dioxochromiumbis(olate)
Synonyms: lead chromate lead(II) chromate
Molecular Weight (g/mol): 323.2
Molecular Formula: CrO4Pb
InChi Key: MOUPNEIJQCETIW-UHFFFAOYSA-N
SMILES: [Pb++].[O-][Cr]([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)