Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Chromate, Powder, ACS, 98%, Spectrum™ Chemical
SDP

Catalog No. 18611227 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
50 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-227 50 kg Poly Bottle
18-611-225 125 g Poly Bottle
18-611-226 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-611-227 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L105550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7758-97-6

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Molecular Weight (g/mol) 323.20
Percent Purity 98%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Lead Chromate, Powder

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.