accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOSOOFFFFFFFFFFFFFFFFF

CAS RN 77620-64-5

IUPAC Name: 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-[(oxiran-2-yl)methyl]-N-propyloctane-1-sulfonamide
Synonyms: 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(oxiran-2-ylmethyl)-N-propyloctane-1-sulfonamide
Molecular Weight (g/mol): 597.29
Molecular Formula: C14H12F17NO3S
InChi Key: HEAWIZXIWLKJKG-UHFFFAOYNA-N
SMILES: CCCN(CC1CO1)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results