accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHOOCH3OOCH3OOCH3OOH3CO

CAS RN 7772-79-4

IUPAC Name: [3,4,6-tris(acetyloxy)-5-acetamidooxan-2-yl]methyl acetate
Synonyms: [3,4,6-tris(acetyloxy)-5-acetamidooxan-2-yl]methyl acetate
Molecular Weight (g/mol): 389.36
Molecular Formula: C16H23NO10
InChi Key: OVPIZHVSWNOZMN-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(OC(C)=O)OC(COC(C)=O)C(OC(C)=O)C1OC(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results